Assay Details
| Gene Symbol: TARID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109982.1 | 7 - 8 | 2367 | 137 | ||
| GenBank mRNA | BC041459.1 | - | 7 - 8 | 2343 | 137 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 326 | LINC00326 | NCRNA00326 | -399.5kb | Anti-sense | Chr. 6 - 133088080 - 133106578 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1312 | LINC01312 | 313.8kb | Anti-sense | Chr. 6 - 133821147 - 133853992 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000419627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000419627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000419627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000451017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000451017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000451017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000451017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000451017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000451017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000451017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000451017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000606544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000607641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000626104.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000626104.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000626104.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000626104.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000626104.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000626104.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000626104.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630119.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630119.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630119.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630119.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630119.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630119.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630119.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630119.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000630728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000631362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000631362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000631362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000631362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000631362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | ENST00000631362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | NR_109982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | NR_109982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | NR_109982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | NR_109982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | NR_109982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | NR_109982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | NR_109982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | NR_109982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | NR_109982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | NR_109982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | NR_109982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | NR_109982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013220.hg.1 | NR_109982.1 | Refseq |

TARID
Genomic Map