Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
ENST00000608258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
ENST00000608258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
ENST00000608258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
ENST00000608258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
ENST00000608258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007411.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
ENST00000608258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
ENST00000608258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
ENST00000608258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
ENST00000608258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
ENST00000608258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007411.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000151289.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151289.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000151289.hg.1 |
TC2000007411.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000151289.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151289.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000151290.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151290.hg.1 |
NR_110004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000151290.hg.1 |
TC2000007411.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000151290.hg.1 |
ENST00000457704.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151290.hg.1 |
NR_110004.1 |
Refseq |