Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
ENST00000419463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
ENST00000419463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
ENST00000419463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
ENST00000419463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
ENST00000419463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
ENST00000419463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006798.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146992.hg.1 |
ENST00000419463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146992.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146992.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146992.hg.1 |
ENST00000419463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146992.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146992.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146992.hg.1 |
TC0700006798.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700146993.hg.1 |
ENST00000419463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146993.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146993.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146993.hg.1 |
ENST00000419463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146993.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146993.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146993.hg.1 |
TC0700006798.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700146994.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146994.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146994.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146994.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146994.hg.1 |
TC0700006798.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700146995.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146995.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146995.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146995.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146995.hg.1 |
TC0700006798.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
ENST00000419463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
ENST00000419463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
ENST00000419463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
ENST00000419463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
ENST00000419463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
ENST00000419463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
NR_110014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
NR_110015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006798.hg.1 |
NR_110015.1 |
Refseq |