Assay Details
Gene Symbol: LOC101926943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110044.1 | 1 - 2 | 212 | 99 | ||
GenBank mRNA | DA836308.1 | - | 1 - 2 | 212 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | GTF2I repeat domain containing 1 | GTF2IRD1 |
BEN, CREAM1, GTF3, MUSTRD1, RBAP2, WBS, WBSCR11, WBSCR12, hMusTRD1alpha1 |
-124.8kb | Anti-sense | Chr. 7 - 74453790 - 74603070 on NCBI Build GRCh38 |
Downstream | neutrophil cytosolic factor 1 | NCF1 |
NCF1A, NOXO2, SH3PXD1A, p47phox |
45.0kb | Anti-sense | Chr. 7 - 74773962 - 74789376 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000595409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000595409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000595409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011516.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006435.hg.1 | ENST00000527297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006435.hg.1 | ENST00000527297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006435.hg.1 | ENST00000527297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011308.hg.1 | ENST00000563515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011308.hg.1 | ENST00000563515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011308.hg.1 | ENST00000565965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011308.hg.1 | ENST00000565965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011308.hg.1 | ENST00000565965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011308.hg.1 | ENST00000565965.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181883.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181883.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181883.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181883.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181883.hg.1 | TC0700011516.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600011308.hg.1 | ENST00000563515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011308.hg.1 | ENST00000563515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011308.hg.1 | ENST00000565965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011308.hg.1 | ENST00000565965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011308.hg.1 | ENST00000565965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011308.hg.1 | ENST00000565965.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181880.hg.1 | TC0700011516.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700181881.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181881.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181881.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181881.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181881.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181881.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181881.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181881.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181881.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181881.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181881.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181881.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181881.hg.1 | TC0700011516.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100006435.hg.1 | ENST00000527297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006435.hg.1 | ENST00000527297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006435.hg.1 | ENST00000527297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000595409.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000595409.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181877.hg.1 | TC0700011516.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700181882.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181882.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181882.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181882.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181882.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181882.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181882.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181882.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181882.hg.1 | TC0700011516.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000434256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000450426.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000594967.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000595409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000595409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000595409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000597786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000601921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | ENST00000619652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | NR_110044.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011516.hg.1 | NR_110044.1 | Refseq |
