Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
ENST00000549844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
ENST00000549844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
ENST00000549844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
ENST00000549844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
ENST00000549844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
ENST00000549844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
ENST00000550431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
ENST00000550431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
ENST00000550431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006842.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147542.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147542.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147542.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147542.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147542.hg.1 |
TC1400006842.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
ENST00000549844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
ENST00000549844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
ENST00000549844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
ENST00000549844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
ENST00000549844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
ENST00000549844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
ENST00000550431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
ENST00000550431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
ENST00000550431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006842.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147539.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147539.hg.1 |
ENST00000549844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147539.hg.1 |
ENST00000549844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147539.hg.1 |
ENST00000550431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147539.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147539.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147539.hg.1 |
ENST00000549844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147539.hg.1 |
ENST00000549844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147539.hg.1 |
ENST00000550431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147539.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147539.hg.1 |
TC1400006842.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400147536.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147536.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147536.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147536.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147536.hg.1 |
TC1400006842.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400147537.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147537.hg.1 |
ENST00000550431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147537.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147537.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147537.hg.1 |
ENST00000550431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147537.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147537.hg.1 |
TC1400006842.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400147543.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147543.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147543.hg.1 |
ENST00000502430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400147543.hg.1 |
NR_110045.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400147543.hg.1 |
TC1400006842.hg.1 |
Clariom D TC |