Assay Details
| Gene Symbol: LINC01441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110064.1 | 1 - 2 | 136 | 155 | ||
| GenBank mRNA | AK056050.1 | - | 1 - 2 | 136 | 155 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1440 | LINC01440 | Anti-sense | Chr. 20 - 55423043 - 55426791 on NCBI Build GRCh38 | ||
| Downstream | cerebellin 4 precursor | CBLN4 | CBLNL1 | 570.2kb | Sense | Chr. 20 - 55997357 - 56005472 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01441
Genomic Map