Assay Details
Gene Symbol: LOC101927021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110069.1 | 2 - 3 | 441 | 67 | ||
GenBank mRNA | HY003139.1 | - | 2 - 3 | 265 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger AN1-type containing 2A | ZFAND2A | AIRAP | -2.1kb | Anti-sense | Chr. 7 - 1148850 - 1160219 on NCBI Build GRCh38 |
Downstream | UNC homeobox | UNCX | UNCX4.1 | 67.1kb | Sense | Chr. 7 - 1233018 - 1236977 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700144562.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700144562.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700144562.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700144562.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700144562.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700144562.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700144562.hg.1 | TC0700006493.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
