| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011752.hg.1 |
ENST00000482790.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
ENST00000500016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
ENST00000500016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
ENST00000500016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
ENST00000500016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
ENST00000500016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
ENST00000500016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
ENST00000500016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
NR_110071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
NR_110071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
NR_110071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
NR_110071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
NR_110071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
NR_110071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010769.hg.1 |
NR_110071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007583.hg.1 |
ENST00000473332.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007583.hg.1 |
ENST00000473332.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011752.hg.1 |
ENST00000482790.3 |
Ensembl |