Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
ENST00000446000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
ENST00000446000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
ENST00000446000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
ENST00000446000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008224.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
ENST00000446000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
ENST00000446000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
ENST00000446000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
ENST00000446000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008224.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157079.hg.1 |
ENST00000446000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157079.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157079.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157079.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157079.hg.1 |
ENST00000446000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157079.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157079.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157079.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157079.hg.1 |
TC0700008224.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700157077.hg.1 |
ENST00000446000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157077.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157077.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157077.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157077.hg.1 |
ENST00000446000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157077.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157077.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157077.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157077.hg.1 |
TC0700008224.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700157081.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157081.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157081.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157081.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157081.hg.1 |
TC0700008224.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700157082.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157082.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157082.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157082.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157082.hg.1 |
TC0700008224.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700157080.hg.1 |
ENST00000446000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157080.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157080.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157080.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157080.hg.1 |
ENST00000446000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157080.hg.1 |
ENST00000450977.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157080.hg.1 |
NR_110079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157080.hg.1 |
NR_110080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157080.hg.1 |
TC0700008224.hg.1 |
Clariom D TC |