Assay Details
Gene Symbol: LOC101928449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110089.1 | 2 - 3 | 667 | 124 | ||
GenBank mRNA | BC031963.1 | - | 2 - 3 | 667 | 124 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | PTPRF interacting protein alpha 2 | PPFIA2 | -194.7kb | Anti-sense | Chr. 12 - 81257975 - 81759748 on NCBI Build GRCh38 | |
Downstream | coiled-coil domain containing 59 | CCDC59 |
BR22, HSPC128, TAP26 |
359.2kb | Anti-sense | Chr. 12 - 82352304 - 82358805 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
