| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013412.hg.1 |
ENST00000468960.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013412.hg.1 |
ENST00000468960.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013412.hg.1 |
ENST00000482799.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013412.hg.1 |
ENST00000482799.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013412.hg.1 |
ENST00000482799.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013412.hg.1 |
ENST00000468960.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013412.hg.1 |
ENST00000468960.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013412.hg.1 |
ENST00000482799.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013412.hg.1 |
ENST00000482799.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013412.hg.1 |
ENST00000482799.2 |
Ensembl |