Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197792.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197792.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197792.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197792.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197792.hg.1 |
TC0700013411.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700197785.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197785.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197785.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197785.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197785.hg.1 |
TC0700013411.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700197789.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197789.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197789.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197789.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197789.hg.1 |
TC0700013411.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700197790.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197790.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197790.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197790.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197790.hg.1 |
TC0700013411.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700197786.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197786.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197786.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197786.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197786.hg.1 |
TC0700013411.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013411.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013411.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197791.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197791.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197791.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197791.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197791.hg.1 |
TC0700013411.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700197788.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197788.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197788.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197788.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197788.hg.1 |
TC0700013411.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700197787.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197787.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197787.hg.1 |
ENST00000360902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197787.hg.1 |
NR_110102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197787.hg.1 |
TC0700013411.hg.1 |
Clariom D TC |