Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
ENST00000553921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
ENST00000553921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
ENST00000553921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
ENST00000553921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
ENST00000553921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
NR_110121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
NR_110121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
NR_110121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009917.hg.1 |
NR_110121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
ENST00000553921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
ENST00000553921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
ENST00000553921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
ENST00000553921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
ENST00000553921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
NR_110121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
NR_110121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
NR_110121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009917.hg.1 |
NR_110121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400173045.hg.1 |
ENST00000553921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400173045.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400173045.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400173045.hg.1 |
NR_110121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400173045.hg.1 |
ENST00000553921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400173045.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400173045.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400173045.hg.1 |
NR_110121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400173045.hg.1 |
TC1400009917.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400173047.hg.1 |
ENST00000553921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400173047.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400173047.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400173047.hg.1 |
NR_110121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400173047.hg.1 |
ENST00000553921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400173047.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400173047.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400173047.hg.1 |
NR_110121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400173047.hg.1 |
TC1400009917.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400173048.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400173048.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400173048.hg.1 |
NR_110121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400173048.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400173048.hg.1 |
ENST00000557631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400173048.hg.1 |
NR_110121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400173048.hg.1 |
TC1400009917.hg.1 |
Clariom D TC |