Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008195.hg.1 |
ENST00000550723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008195.hg.1 |
ENST00000550723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008195.hg.1 |
ENST00000550723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008195.hg.1 |
ENST00000550723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008195.hg.1 |
NR_110130.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008195.hg.1 |
NR_110130.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008195.hg.1 |
NR_110130.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008195.hg.1 |
NR_110130.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200160323.hg.1 |
ENST00000550723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200160323.hg.1 |
NR_110130.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200160323.hg.1 |
ENST00000550723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200160323.hg.1 |
NR_110130.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200160323.hg.1 |
TC1200008195.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200008195.hg.1 |
ENST00000550723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008195.hg.1 |
ENST00000550723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008195.hg.1 |
ENST00000550723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008195.hg.1 |
ENST00000550723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008195.hg.1 |
NR_110130.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008195.hg.1 |
NR_110130.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008195.hg.1 |
NR_110130.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008195.hg.1 |
NR_110130.1 |
Refseq |