| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000414856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000443319.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000443319.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000443319.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
ENST00000443319.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198674.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100198674.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100198674.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198674.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100198674.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100198674.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198674.hg.1 |
TC1100012866.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000414856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000443319.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000443319.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000443319.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
ENST00000443319.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012866.hg.1 |
NR_110140.1 |
Refseq |