Assay Details
| Gene Symbol: LOC101929653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110140.1 | 3 - 4 | 295 | 71 | ||
| GenBank mRNA | AK128059.1 | - | 3 - 4 | 295 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sorting nexin 19 | SNX19 | CHET8 | -622.6kb | Sense | Chr. 11 - 130866254 - 130916488 on NCBI Build GRCh38 |
| Downstream | NTM antisense RNA 1 | NTM-AS1 | C11orf39 | 122.5kb | Sense | Chr. 11 - 131662129 - 131663556 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000414856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000443319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000443319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000443319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | ENST00000443319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198673.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198673.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198673.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198673.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198673.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198673.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198673.hg.1 | TC1100012866.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100198672.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198672.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198672.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198672.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198672.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198672.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198672.hg.1 | TC1100012866.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100198671.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198671.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198671.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198671.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198671.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198671.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198671.hg.1 | TC1100012866.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000414856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000443319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000443319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000443319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | ENST00000443319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012866.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198670.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198670.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198670.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198670.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198670.hg.1 | ENST00000414466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198670.hg.1 | NR_110140.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198670.hg.1 | TC1100012866.hg.1 | Clariom D TC |

LOC101929653
Genomic Map