Assay Details
| Gene Symbol: ID2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110154.1 | 2 - 3 | 305 | 99 | ||
| GenBank mRNA | DA317685.1 | - | 2 - 3 | 305 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929567 | LOC101929567 | -87.8kb | Sense | Chr. 2 - 8559833 - 8583792 on NCBI Build GRCh38 | |
| Downstream |
inhibitor of DNA binding 2, HLH protein |
ID2 |
GIG8, ID2A, ID2H, bHLHb26 |
8.7kb | Anti-sense | Chr. 2 - 8681983 - 8684453 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000418957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000418957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000418957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000418957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000421298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000421298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000433340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000433340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000433340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000433592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000433592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000433592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000433592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | NR_110153.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | NR_110153.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184703.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184703.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184703.hg.1 | NR_110153.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184703.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184703.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184703.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184703.hg.1 | NR_110153.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184703.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184703.hg.1 | TC0200011664.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200184683.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184683.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184683.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184683.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184683.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184683.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184683.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184683.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184683.hg.1 | TC0200011664.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200184684.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184684.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184684.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184684.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184684.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184684.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184684.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184684.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184684.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184684.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184684.hg.1 | TC0200011664.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200184682.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184682.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184682.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184682.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184682.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184682.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184682.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184682.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184682.hg.1 | TC0200011664.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000418957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000418957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000418957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000418957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000421298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000421298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000433340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000433340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000433340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000433592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000433592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000433592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000433592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | NR_110153.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | NR_110153.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | NR_110154.1 | Refseq |

ID2-AS1
Genomic Map