Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000499910.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000499910.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000499910.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000499910.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000499910.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000499910.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000499910.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000499910.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000554260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000554260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000554260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000554260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000554260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000554260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000554260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000554862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000554862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000554862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000554862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000554862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000554862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
ENST00000554862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
NR_110166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
NR_110166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
NR_110166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
NR_110166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
NR_110166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008141.hg.1 |
NR_110166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000499910.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000499910.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000499910.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000499910.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000499910.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000499910.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000499910.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000499910.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000554260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000554260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000554260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000554260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000554260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000554260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000554260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000554862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000554862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000554862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000554862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000554862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000554862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
ENST00000554862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
NR_110165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
NR_110166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
NR_110166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
NR_110166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
NR_110166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
NR_110166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008141.hg.1 |
NR_110166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400157944.hg.1 |
ENST00000554862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400157944.hg.1 |
NR_110166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400157944.hg.1 |
ENST00000554862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400157944.hg.1 |
NR_110166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400157944.hg.1 |
TC1400008141.hg.1 |
Clariom D TC |