Assay Details
| Gene Symbol: U2AF1L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110171.1 | 2 - 3 | 160 | 115 | ||
| NR_110172.1 | 2 - 3 | 160 | 115 | |||
| NR_110173.1 | 2 - 3 | 185 | 115 | |||
| GenBank mRNA | AY461582.1 | - | 2 - 3 | 134 | 115 | |
| BM696851.1 | - | 2 - 3 | 178 | 115 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | IGF like family receptor 1 | IGFLR1 |
TMEM149, U2AF1L4 |
-2.2kb | Sense | Chr. 19 - 35739250 - 35742450 on NCBI Build GRCh38 |
| Downstream | presenilin enhancer gamma-secretase subunit | PSENEN |
MDS033, MSTP064, PEN-2, PEN2 |
390.0 | Anti-sense | Chr. 19 - 35745577 - 35747155 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

U2AF1L4
Genomic Map