Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
ENST00000420579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
ENST00000420579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
ENST00000420579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
ENST00000420579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
ENST00000420579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
ENST00000435407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
ENST00000435407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
ENST00000435407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
ENST00000446401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
ENST00000446401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
ENST00000446401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
ENST00000446401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
ENST00000602760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
ENST00000602760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013949.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
ENST00000420579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
ENST00000420579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
ENST00000420579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
ENST00000420579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
ENST00000420579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
ENST00000435407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
ENST00000435407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
ENST00000435407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
ENST00000446401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
ENST00000446401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
ENST00000446401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
ENST00000446401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
ENST00000602760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
ENST00000602760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013949.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202357.hg.1 |
ENST00000420579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202357.hg.1 |
ENST00000446401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202357.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202357.hg.1 |
ENST00000420579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202357.hg.1 |
ENST00000446401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202357.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202357.hg.1 |
TC0200013949.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200202362.hg.1 |
ENST00000446401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202362.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202362.hg.1 |
ENST00000446401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202362.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202362.hg.1 |
TC0200013949.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200202384.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202384.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202384.hg.1 |
TC0200013949.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200202371.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202371.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202371.hg.1 |
TC0200013949.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200202363.hg.1 |
ENST00000446401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202363.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202363.hg.1 |
ENST00000446401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202363.hg.1 |
NR_110174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202363.hg.1 |
TC0200013949.hg.1 |
Clariom D TC |