Assay Details
| Gene Symbol: MIR3681HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110196.1 | 1 - 2 | 113 | 97 | ||
| NR_110197.1 | 1 - 2 | 113 | 97 | |||
| NR_110198.1 | 1 - 2 | 113 | 97 | |||
| GenBank mRNA | AK001558.1 | - | 1 - 2 | 101 | 97 | |
| BX439624.2 | - | 1 - 2 | 112 | 97 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4262 | MIR4262 | -170.1kb | Anti-sense | Chr. 2 - 11836933 - 11836986 on NCBI Build GRCh38 | |
| Downstream | microRNA 3681 | MIR3681 | 191.3kb | Sense | Chr. 2 - 12199130 - 12199201 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

MIR3681HG
Genomic Map