Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
ENST00000436967.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
ENST00000436967.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
ENST00000436967.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
ENST00000436967.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
ENST00000476839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
ENST00000476839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
ENST00000476839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
ENST00000476839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006790.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146567.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146567.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146567.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146567.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146567.hg.1 |
TC0200006790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
ENST00000436967.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
ENST00000436967.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
ENST00000436967.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
ENST00000436967.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
ENST00000476839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
ENST00000476839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
ENST00000476839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
ENST00000476839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006790.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146568.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146568.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146568.hg.1 |
ENST00000492352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146568.hg.1 |
NR_110201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146568.hg.1 |
TC0200006790.hg.1 |
Clariom D TC |