Assay Details
| Gene Symbol: LOC101927043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110208.1 | 3 - 4 | 405 | 85 | ||
| GenBank mRNA | DA483349.1 | - | 3 - 4 | 406 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calmodulin 2 | CALM2 |
CAMII, LQT15, PHKD, PHKD2, caM |
-22.8kb | Sense | Chr. 2 - 47160082 - 47176936 on NCBI Build GRCh38 |
| Downstream | brain cytoplasmic RNA 1 | BCYRN1 |
BC200, BC200a, LINC00004, NCRNA00004 |
2.9kb | Anti-sense | Chr. 2 - 47335315 - 47335514 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191313.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191313.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191313.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191313.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191313.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191313.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191313.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191313.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191313.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191313.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191313.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191313.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191313.hg.1 | TC0200012505.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191301.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191301.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191301.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191301.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191301.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191301.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191301.hg.1 | TC0200012505.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200191318.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191318.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191318.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191318.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191318.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191318.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191318.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191318.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191318.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191318.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191318.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191318.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191318.hg.1 | TC0200012505.hg.1 | Clariom D TC |

LOC101927043
Genomic Map