Assay Details
| Gene Symbol: LOC101927043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110208.1 | 4 - 5 | 515 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191300.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191300.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191300.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191300.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191300.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191300.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191300.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191300.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191300.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191300.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191300.hg.1 | TC0200012505.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200191299.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191299.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191299.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191299.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191299.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200191299.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200191299.hg.1 | TC0200012505.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000413185.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000419035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000441997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000448713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | ENST00000450550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012505.hg.1 | NR_110208.1 | Refseq |

LOC101927043
Genomic Map