| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000419719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000419719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000419719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000419719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000419719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
NR_110218.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
NR_110218.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
NR_110218.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
NR_110218.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016758.hg.1 |
NR_110218.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200228673.hg.1 |
ENST00000419719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200228673.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200228673.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200228673.hg.1 |
NR_110218.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200228673.hg.1 |
ENST00000419719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200228673.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200228673.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200228673.hg.1 |
NR_110218.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200228673.hg.1 |
TC0200016758.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000419719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000419719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000419719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000419719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000419719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000427108.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
ENST00000456653.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
NR_110218.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
NR_110218.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
NR_110218.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
NR_110218.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016758.hg.1 |
NR_110218.1 |
Refseq |