Assay Details
Gene Symbol: LOC101927438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110224.1 | 3 - 4 | 415 | 95 | ||
GenBank mRNA | AK097952.1 | - | 3 - 4 | 410 | 95 | |
AV712717.1 | - | 3 - 4 | 414 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SERTA domain containing 2 | SERTAD2 |
Sei-2, TRIP-Br2 |
-96.7kb | Sense | Chr. 2 - 64631621 - 64751091 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC400958 | LOC400958 | 53.5kb | Sense | Chr. 2 - 64901840 - 64932447 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
