Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014669.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206687.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206687.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206687.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206687.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206687.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206687.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206687.hg.1 |
TC0200014669.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200206685.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206685.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206685.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206685.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206685.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206685.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206685.hg.1 |
TC0200014669.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200206691.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206691.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206691.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206691.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206691.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206691.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206691.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206691.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206691.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206691.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206691.hg.1 |
TC0200014669.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
ENST00000448255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014669.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206686.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206686.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206686.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206686.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206686.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206686.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206686.hg.1 |
TC0200014669.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200206688.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206688.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206688.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206688.hg.1 |
ENST00000428651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206688.hg.1 |
NR_110249.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206688.hg.1 |
NR_110250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206688.hg.1 |
TC0200014669.hg.1 |
Clariom D TC |