Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000413227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000413227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000437979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000437979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000437979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226104.hg.1 |
ENST00000413227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226104.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226104.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226104.hg.1 |
ENST00000437979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226104.hg.1 |
NR_110259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226104.hg.1 |
ENST00000413227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226104.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226104.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226104.hg.1 |
ENST00000437979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226104.hg.1 |
NR_110259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226104.hg.1 |
TC0200016653.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000413227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000413227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000437979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000437979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000437979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |