Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015357.hg.1 |
ENST00000443261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015357.hg.1 |
ENST00000443261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015357.hg.1 |
ENST00000443261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015357.hg.1 |
ENST00000451266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015357.hg.1 |
ENST00000451266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015357.hg.1 |
ENST00000456248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015357.hg.1 |
ENST00000456248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015357.hg.1 |
ENST00000456248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015357.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015357.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015357.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015357.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015357.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212556.hg.1 |
ENST00000443261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212556.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212556.hg.1 |
ENST00000443261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212556.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212556.hg.1 |
TC0200015357.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200212551.hg.1 |
ENST00000456248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212551.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212551.hg.1 |
ENST00000456248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212551.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212551.hg.1 |
TC0200015357.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200015357.hg.1 |
ENST00000443261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015357.hg.1 |
ENST00000443261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015357.hg.1 |
ENST00000443261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015357.hg.1 |
ENST00000451266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015357.hg.1 |
ENST00000451266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015357.hg.1 |
ENST00000456248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015357.hg.1 |
ENST00000456248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015357.hg.1 |
ENST00000456248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015357.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015357.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015357.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015357.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015357.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212552.hg.1 |
ENST00000451266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212552.hg.1 |
ENST00000456248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212552.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212552.hg.1 |
ENST00000451266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212552.hg.1 |
ENST00000456248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212552.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212552.hg.1 |
TC0200015357.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200212555.hg.1 |
ENST00000443261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212555.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212555.hg.1 |
ENST00000443261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212555.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212555.hg.1 |
TC0200015357.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200212554.hg.1 |
ENST00000443261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212554.hg.1 |
ENST00000456248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212554.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212554.hg.1 |
ENST00000443261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212554.hg.1 |
ENST00000456248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212554.hg.1 |
NR_110267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212554.hg.1 |
TC0200015357.hg.1 |
Clariom D TC |