Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016685.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016685.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016685.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016685.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016685.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016685.hg.1 |
ENST00000447639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016685.hg.1 |
ENST00000447639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016685.hg.1 |
ENST00000447639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016685.hg.1 |
NR_110273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016685.hg.1 |
NR_110273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016685.hg.1 |
NR_110273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016685.hg.1 |
NR_110273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016685.hg.1 |
NR_110274.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016685.hg.1 |
NR_110274.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016685.hg.1 |
NR_110274.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016685.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016685.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016685.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016685.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016685.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016685.hg.1 |
ENST00000447639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016685.hg.1 |
ENST00000447639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016685.hg.1 |
ENST00000447639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016685.hg.1 |
NR_110273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016685.hg.1 |
NR_110273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016685.hg.1 |
NR_110273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016685.hg.1 |
NR_110273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016685.hg.1 |
NR_110274.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016685.hg.1 |
NR_110274.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016685.hg.1 |
NR_110274.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226702.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226702.hg.1 |
NR_110273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226702.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226702.hg.1 |
NR_110273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226702.hg.1 |
TC0200016685.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200226703.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226703.hg.1 |
ENST00000447639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226703.hg.1 |
NR_110273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226703.hg.1 |
NR_110274.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226703.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226703.hg.1 |
ENST00000447639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226703.hg.1 |
NR_110273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226703.hg.1 |
NR_110274.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226703.hg.1 |
TC0200016685.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200226704.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226704.hg.1 |
NR_110273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226704.hg.1 |
NR_110274.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226704.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226704.hg.1 |
NR_110273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226704.hg.1 |
NR_110274.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226704.hg.1 |
TC0200016685.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200226701.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226701.hg.1 |
ENST00000447639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226701.hg.1 |
NR_110273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226701.hg.1 |
NR_110274.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226701.hg.1 |
ENST00000422761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226701.hg.1 |
ENST00000447639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226701.hg.1 |
NR_110273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226701.hg.1 |
NR_110274.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226701.hg.1 |
TC0200016685.hg.1 |
Clariom D TC |