Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013181.hg.1 |
ENST00000443301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013181.hg.1 |
ENST00000443301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013181.hg.1 |
ENST00000443301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013181.hg.1 |
ENST00000443301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013181.hg.1 |
ENST00000443301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013181.hg.1 |
NR_110289.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013181.hg.1 |
NR_110289.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013181.hg.1 |
NR_110289.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013181.hg.1 |
NR_110289.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013181.hg.1 |
NR_110289.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200196470.hg.1 |
ENST00000443301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196470.hg.1 |
NR_110289.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200196470.hg.1 |
ENST00000443301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196470.hg.1 |
NR_110289.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200196470.hg.1 |
TC0200013181.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200196471.hg.1 |
ENST00000443301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196471.hg.1 |
NR_110289.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200196471.hg.1 |
ENST00000443301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196471.hg.1 |
NR_110289.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200196471.hg.1 |
TC0200013181.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200013181.hg.1 |
ENST00000443301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013181.hg.1 |
ENST00000443301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013181.hg.1 |
ENST00000443301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013181.hg.1 |
ENST00000443301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013181.hg.1 |
ENST00000443301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013181.hg.1 |
NR_110289.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013181.hg.1 |
NR_110289.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013181.hg.1 |
NR_110289.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013181.hg.1 |
NR_110289.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013181.hg.1 |
NR_110289.1 |
Refseq |