Assay Details
Gene Symbol: LOC101928020 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110291.1 | 1 - 2 | 74 | 110 | ||
GenBank mRNA | AW168234.1 | - | 2 - 3 | 110 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | KAT8 regulatory NSL complex subunit 1 like | KANSL1L |
C2orf67, MSL1v2 |
Anti-sense | Chr. 2 - 210021421 - 210171370 on NCBI Build GRCh38 | |
Downstream |
acyl-CoA dehydrogenase, long chain |
ACADL |
ACAD4, LCAD |
156.5kb | Anti-sense | Chr. 2 - 210187990 - 210225491 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
