Assay Details
| Gene Symbol: LINC01246 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110498.1 | 2 - 3 | 418 | 135 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1247 | LINC01247 | -250.7kb | Sense | Chr. 2 - 6366010 - 6375422 on NCBI Build GRCh38 | |
| Downstream | MIR7515 host gene | MIR7515HG | 15.1kb | Sense | Chr. 2 - 6649107 - 6650519 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000456028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000456028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000585834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000585834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000585872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000585872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000586753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000586753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000587316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000587316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000588384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000588384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000589024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000589597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000589597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000589962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000589962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000590276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000590276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000590489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000590489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000590865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000590865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000591018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000591018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000591053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000591053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000591060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000591060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000591450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000591450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000592394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000592394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000592565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000592565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000593202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000593202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000625915.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000626844.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | NR_110498.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000456028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000456028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000585834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000585834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000585872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000585872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000586753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000586753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000587316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000587316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000588384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000588384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000589024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000589597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000589597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000589962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000589962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000590276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000590276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000590489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000590489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000590865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000590865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000591018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000591018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000591053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000591053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000591060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000591060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000591450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000591450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000592394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000592394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000592565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000592565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000593202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000593202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000625915.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | ENST00000626844.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | NR_110498.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184262.hg.1 | TC0200011617.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000455317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000455317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000456028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000456028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000585834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000585834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000587748.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000587748.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000589597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000589597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000590276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000590276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000590971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000590971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000591060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000591060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000591575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000591575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000593202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000593202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000625915.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000629313.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | NR_110498.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | NR_110500.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000455317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000455317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000456028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000456028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000585834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000585834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000587748.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000587748.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000589597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000589597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000590276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000590276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000590971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000590971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000591060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000591060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000591575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000591575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000593202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000593202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000625915.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | ENST00000629313.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | NR_110498.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | NR_110500.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184249.hg.1 | TC0200011617.hg.1 | Clariom D TC |

LINC01246
Genomic Map