Assay Details
| Gene Symbol: CHKB-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110536.1 | 2 - 3 | 403 | 79 | ||
| GenBank mRNA | BI561134.1 | - | 2 - 3 | 310 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CHKB-CPT1B readthrough (NMD candidate) | CHKB-CPT1B | CHKL-CPT1B | -3.8kb | Anti-sense | Chr. 22 - 50568861 - 50582999 on NCBI Build GRCh38 |
| Downstream | mitogen-activated protein kinase 8 interacting protein 2 | MAPK8IP2 |
IB-2, IB2, JIP2, PRKM8IPL |
13.2kb | Sense | Chr. 22 - 50600685 - 50613978 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

CHKB-AS1
Genomic Map