Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006683.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006683.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006683.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006683.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006683.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006683.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006683.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006683.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006683.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006683.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145974.hg.1 |
TC2200006683.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200145974.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145974.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145974.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145974.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145970.hg.1 |
TC2200006683.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200145970.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145970.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145970.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145970.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145973.hg.1 |
TC2200006683.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200145973.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145973.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145973.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145973.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145971.hg.1 |
TC2200006683.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200145971.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145971.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145971.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145971.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145972.hg.1 |
TC2200006683.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200145972.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145972.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145972.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145972.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006683.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006683.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006683.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006683.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006683.hg.1 |
ENST00000444039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006683.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006683.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006683.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006683.hg.1 |
NR_110537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006683.hg.1 |
NR_110537.1 |
Refseq |