Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000548104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000548104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000548104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000548361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000548361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
ENST00000548361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160712.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160712.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160712.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160712.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160712.hg.1 |
TC1400008469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400160706.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160706.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160706.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160706.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160706.hg.1 |
TC1400008469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400160707.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160707.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160707.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160707.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160707.hg.1 |
TC1400008469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400160716.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160716.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160716.hg.1 |
TC1400008469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000548104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000548104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000548104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000548361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000548361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
ENST00000548361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008469.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160711.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160711.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160711.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160711.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160711.hg.1 |
TC1400008469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400160701.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160701.hg.1 |
ENST00000548361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160701.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160701.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160701.hg.1 |
ENST00000548361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160701.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160701.hg.1 |
TC1400008469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400160710.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160710.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160710.hg.1 |
ENST00000546968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160710.hg.1 |
NR_110546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160710.hg.1 |
TC1400008469.hg.1 |
Clariom D TC |