| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000553762.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000553762.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000553762.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000553762.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000553762.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000553762.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556026.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556026.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556026.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556026.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556026.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
NR_110547.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
NR_110547.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
NR_110547.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
NR_110547.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
NR_110547.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
NR_110547.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007323.hg.1 |
NR_110547.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150855.hg.1 |
ENST00000556026.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150855.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150855.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150855.hg.1 |
NR_110547.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150855.hg.1 |
ENST00000556026.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150855.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150855.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150855.hg.1 |
NR_110547.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150855.hg.1 |
TC1400007323.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000553762.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000553762.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000553762.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000553762.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000553762.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000553762.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000555378.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556026.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556026.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556026.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556026.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556026.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
ENST00000556815.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
NR_110547.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
NR_110547.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
NR_110547.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
NR_110547.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
NR_110547.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
NR_110547.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007323.hg.1 |
NR_110547.1 |
Refseq |