Assay Details
| Gene Symbol: AANAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110548.1 | 2 - 3 | 415 | 105 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin conjugating enzyme E2 O | UBE2O | E2-230K | -15.5kb | Anti-sense | Chr. 17 - 76389532 - 76453206 on NCBI Build GRCh38 |
| Downstream | rhomboid 5 homolog 2 | RHBDF2 |
RHBDL5, RHBDL6, TEC, TOC, TOCG, iRhom2 |
1.6kb | Anti-sense | Chr. 17 - 76470893 - 76501440 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
N-terminal protein amino acid acetylation
circadian rhythm
photoperiodism
response to zinc ion
melatonin biosynthetic process
response to insulin
response to cytokine
response to prostaglandin E
indolalkylamine biosynthetic process
response to copper ion
response to corticosterone
response to calcium ion
cellular response to cAMP
aralkylamine N-acetyltransferase activity
arylamine N-acetyltransferase activity
protein binding
14-3-3 protein binding
N-terminal protein amino acid acetylation
circadian rhythm
photoperiodism
response to zinc ion
melatonin biosynthetic process
response to insulin
response to cytokine
response to prostaglandin E
indolalkylamine biosynthetic process
response to copper ion
response to corticosterone
response to calcium ion
cellular response to cAMP
aralkylamine N-acetyltransferase activity
arylamine N-acetyltransferase activity
protein binding
14-3-3 protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000392492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000392492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000392492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000392492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000392492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000392492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000392492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000392492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000585649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000585649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000585649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000585649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000587798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000587798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000587798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000587798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | ENST00000587798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001088.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001088.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001088.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001088.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001088.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001088.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001088.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001088.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700167408.hg.1 | ENST00000587798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167408.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700167408.hg.1 | ENST00000587798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167408.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700167408.hg.1 | TC1700008946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000250615.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000392492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000392492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000392492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000392492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000392492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000392492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000392492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000392492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000585649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000585649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000585649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000585649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000587798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000587798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000587798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000587798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | ENST00000587798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001088.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001088.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001088.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001088.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001088.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001088.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001088.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001088.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NM_001166579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NR_110548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008946.hg.1 | NR_110548.1 | Refseq |

AANAT
Genomic Map