Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
ENST00000550214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
ENST00000550214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
ENST00000550214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
ENST00000550214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
ENST00000550530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
ENST00000550530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
ENST00000550530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
ENST00000550530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
ENST00000552379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
ENST00000552379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
ENST00000552379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
ENST00000552379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
NR_110589.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
NR_110589.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
NR_110589.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
NR_110590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
NR_110590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
NR_110590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
NR_110590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
NR_110591.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
NR_110591.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
NR_110591.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010647.hg.1 |
NR_110591.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181060.hg.1 |
ENST00000550214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181060.hg.1 |
ENST00000552379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181060.hg.1 |
NR_110589.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181060.hg.1 |
NR_110590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181060.hg.1 |
ENST00000550214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181060.hg.1 |
ENST00000552379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181060.hg.1 |
NR_110589.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181060.hg.1 |
NR_110590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181060.hg.1 |
TC1200010647.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
ENST00000550214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
ENST00000550214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
ENST00000550214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
ENST00000550214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
ENST00000550530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
ENST00000550530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
ENST00000550530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
ENST00000550530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
ENST00000552379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
ENST00000552379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
ENST00000552379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
ENST00000552379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
NR_110589.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
NR_110589.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
NR_110589.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
NR_110590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
NR_110590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
NR_110590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
NR_110590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
NR_110591.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
NR_110591.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
NR_110591.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010647.hg.1 |
NR_110591.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181056.hg.1 |
ENST00000552379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181056.hg.1 |
NR_110590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181056.hg.1 |
ENST00000552379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181056.hg.1 |
NR_110590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181056.hg.1 |
TC1200010647.hg.1 |
Clariom D TC |