Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011908.hg.1 |
ENST00000417605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011908.hg.1 |
ENST00000417605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011908.hg.1 |
ENST00000417605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011908.hg.1 |
ENST00000417605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011908.hg.1 |
ENST00000417605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011908.hg.1 |
NR_110630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011908.hg.1 |
NR_110630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011908.hg.1 |
NR_110630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011908.hg.1 |
NR_110630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187875.hg.1 |
NR_110630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187875.hg.1 |
NR_110630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187875.hg.1 |
TC0100011908.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100187876.hg.1 |
ENST00000417605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187876.hg.1 |
NR_110630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187876.hg.1 |
ENST00000417605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187876.hg.1 |
NR_110630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187876.hg.1 |
TC0100011908.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100187878.hg.1 |
ENST00000417605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187878.hg.1 |
NR_110630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187878.hg.1 |
ENST00000417605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187878.hg.1 |
NR_110630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187878.hg.1 |
TC0100011908.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100187879.hg.1 |
ENST00000417605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187879.hg.1 |
NR_110630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187879.hg.1 |
ENST00000417605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187879.hg.1 |
NR_110630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187879.hg.1 |
TC0100011908.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011908.hg.1 |
ENST00000417605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011908.hg.1 |
ENST00000417605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011908.hg.1 |
ENST00000417605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011908.hg.1 |
ENST00000417605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011908.hg.1 |
ENST00000417605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011908.hg.1 |
NR_110630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011908.hg.1 |
NR_110630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011908.hg.1 |
NR_110630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011908.hg.1 |
NR_110630.1 |
Refseq |