Assay Details
| Gene Symbol: LINC01230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110643.1 | 1 | 120 | 74 | ||
| Gene Symbol: DMRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011517687.1 | XP_011515989.1 | 2 | 1154 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | doublesex and mab-3 related transcription factor 3 | DMRT3 | DMRTA3 | -56.4kb | Sense | Chr. 9 - 976968 - 991732 on NCBI Build GRCh38 | 
| Downstream | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2 | SMARCA2 | BAF190, BRM, NCBRS, SNF2, SNF2L2, SNF2LA, SWI2, Sth1p, hBRM, hSNF2a | 966.6kb | Sense | Chr. 9 - 2015219 - 2193624 on NCBI Build GRCh38 | 
 Back To Top
 Back To TopMore Information
Updated Mapping Information
 This assay targets the same location as Clariom D transcript cluster(s) TC0900006457.hg.1
					            	This assay targets the same location as Clariom D transcript cluster(s) TC0900006457.hg.1Additional Information:
				    	 
					    	
					        	
					            	This assay is in the 5'UTR region of the XM_011517687.1 transcripts.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Amplicon length between 71 and 85  3' Most  Detect Genomic DNA  Homo sapiens  Both primers and probe map within a single exon | 
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
 Back To Top
 Back To Top
 LINC01230
LINC01230 Genomic Map
Genomic Map Back To Top
 Back To Top