Assay Details
Gene Symbol: ERICH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110644.1 | 3 - 4 | 848 | 105 | ||
GenBank mRNA | BQ269226.1 | - | 3 - 4 | 417 | 105 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101926913 | LOC101926913 | -6.4kb | Anti-sense | Chr. 2 - 170700368 - 170770766 on NCBI Build GRCh38 | |
Downstream | glutamate decarboxylase 1 | GAD1 |
CPSQ1, GAD, SCP |
35.8kb | Sense | Chr. 2 - 170813210 - 170861151 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000409885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001289947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009933.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169564.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169564.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169564.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169564.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169564.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169564.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169564.hg.1 | ENST00000426475.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169564.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169564.hg.1 | ENST00000429172.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200169564.hg.1 | NM_001290030.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169564.hg.1 | NM_001290031.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169564.hg.1 | NR_110644.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200169564.hg.1 | TC0200009933.hg.1 | Clariom D TC |
