Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010207.hg.1 |
ENST00000570024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010207.hg.1 |
ENST00000570024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010207.hg.1 |
NR_110649.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010207.hg.1 |
NR_110649.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178583.hg.1 |
ENST00000570024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178583.hg.1 |
NR_110649.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178583.hg.1 |
ENST00000570024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178583.hg.1 |
NR_110649.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178583.hg.1 |
TC1600010207.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600178584.hg.1 |
ENST00000570024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178584.hg.1 |
NR_110649.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178584.hg.1 |
ENST00000570024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178584.hg.1 |
NR_110649.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178584.hg.1 |
TC1600010207.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600010207.hg.1 |
ENST00000570024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010207.hg.1 |
ENST00000570024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010207.hg.1 |
NR_110649.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010207.hg.1 |
NR_110649.1 |
Refseq |