Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008174.hg.1 |
ENST00000451135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008174.hg.1 |
ENST00000451135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008174.hg.1 |
ENST00000451135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008174.hg.1 |
ENST00000451135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008174.hg.1 |
NR_110658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008174.hg.1 |
NR_110658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008174.hg.1 |
NR_110658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008174.hg.1 |
NR_110658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008174.hg.1 |
NR_110658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157201.hg.1 |
ENST00000451135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157201.hg.1 |
NR_110658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157201.hg.1 |
ENST00000451135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157201.hg.1 |
NR_110658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157201.hg.1 |
TC0600008174.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008174.hg.1 |
ENST00000451135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008174.hg.1 |
ENST00000451135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008174.hg.1 |
ENST00000451135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008174.hg.1 |
ENST00000451135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008174.hg.1 |
NR_110658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008174.hg.1 |
NR_110658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008174.hg.1 |
NR_110658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008174.hg.1 |
NR_110658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008174.hg.1 |
NR_110658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157202.hg.1 |
ENST00000451135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157202.hg.1 |
NR_110658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157202.hg.1 |
ENST00000451135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157202.hg.1 |
NR_110658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157202.hg.1 |
TC0600008174.hg.1 |
Clariom D TC |