Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010729.hg.1 |
ENST00000427173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010729.hg.1 |
ENST00000427173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010729.hg.1 |
ENST00000427173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010729.hg.1 |
ENST00000427173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010729.hg.1 |
NR_110722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010729.hg.1 |
NR_110722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010729.hg.1 |
NR_110722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010729.hg.1 |
NR_110722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178779.hg.1 |
ENST00000427173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178779.hg.1 |
NR_110722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178779.hg.1 |
ENST00000427173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178779.hg.1 |
NR_110722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178779.hg.1 |
TC0100010729.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100178781.hg.1 |
ENST00000427173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178781.hg.1 |
NR_110722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178781.hg.1 |
ENST00000427173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178781.hg.1 |
NR_110722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178781.hg.1 |
TC0100010729.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010729.hg.1 |
ENST00000427173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010729.hg.1 |
ENST00000427173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010729.hg.1 |
ENST00000427173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010729.hg.1 |
ENST00000427173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010729.hg.1 |
NR_110722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010729.hg.1 |
NR_110722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010729.hg.1 |
NR_110722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010729.hg.1 |
NR_110722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178778.hg.1 |
ENST00000427173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178778.hg.1 |
NR_110722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178778.hg.1 |
ENST00000427173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178778.hg.1 |
NR_110722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178778.hg.1 |
TC0100010729.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100178780.hg.1 |
ENST00000427173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178780.hg.1 |
NR_110722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178780.hg.1 |
ENST00000427173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178780.hg.1 |
NR_110722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178780.hg.1 |
TC0100010729.hg.1 |
Clariom D TC |