Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010457.hg.1 |
ENST00000604333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010457.hg.1 |
ENST00000604333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010457.hg.1 |
ENST00000604333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010457.hg.1 |
NR_110749.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010457.hg.1 |
NR_110749.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010457.hg.1 |
NR_110749.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185467.hg.1 |
NR_110749.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185467.hg.1 |
TC1900010457.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900185467.hg.1 |
NR_110749.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185475.hg.1 |
ENST00000604333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185475.hg.1 |
NR_110749.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185475.hg.1 |
TC1900010457.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900185475.hg.1 |
ENST00000604333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185475.hg.1 |
NR_110749.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010457.hg.1 |
ENST00000604333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010457.hg.1 |
ENST00000604333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010457.hg.1 |
ENST00000604333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010457.hg.1 |
NR_110749.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010457.hg.1 |
NR_110749.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010457.hg.1 |
NR_110749.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185470.hg.1 |
ENST00000604333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185470.hg.1 |
NR_110749.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185470.hg.1 |
TC1900010457.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900185470.hg.1 |
ENST00000604333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185470.hg.1 |
NR_110749.1 |
Refseq |