Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011712.hg.1 |
ENST00000453968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011712.hg.1 |
ENST00000453968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011712.hg.1 |
ENST00000453968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011712.hg.1 |
NR_110750.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011712.hg.1 |
NR_110750.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011712.hg.1 |
NR_110750.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900200419.hg.1 |
ENST00000453968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200419.hg.1 |
NR_110750.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900200419.hg.1 |
TC1900011712.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900200419.hg.1 |
ENST00000453968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200419.hg.1 |
NR_110750.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900200421.hg.1 |
ENST00000453968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200421.hg.1 |
NR_110750.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900200421.hg.1 |
TC1900011712.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900200421.hg.1 |
ENST00000453968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200421.hg.1 |
NR_110750.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011712.hg.1 |
ENST00000453968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011712.hg.1 |
ENST00000453968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011712.hg.1 |
ENST00000453968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011712.hg.1 |
NR_110750.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011712.hg.1 |
NR_110750.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011712.hg.1 |
NR_110750.1 |
Refseq |