Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006941.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006941.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006941.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006941.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006941.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006941.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006941.hg.1 |
NR_110762.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006941.hg.1 |
NR_110762.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006941.hg.1 |
NR_110762.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006941.hg.1 |
NR_110762.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006941.hg.1 |
NR_110762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800147698.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800147698.hg.1 |
NR_110762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800147698.hg.1 |
TC1800006941.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800147698.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800147698.hg.1 |
NR_110762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800147699.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800147699.hg.1 |
NR_110762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800147699.hg.1 |
TC1800006941.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800147699.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800147699.hg.1 |
NR_110762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800147700.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800147700.hg.1 |
NR_110762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800147700.hg.1 |
TC1800006941.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800147700.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800147700.hg.1 |
NR_110762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006941.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006941.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006941.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006941.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006941.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006941.hg.1 |
ENST00000578367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006941.hg.1 |
NR_110762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006941.hg.1 |
NR_110762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006941.hg.1 |
NR_110762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006941.hg.1 |
NR_110762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006941.hg.1 |
NR_110762.1 |
Refseq |