Assay Details
Gene Symbol: LINC01543 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110762.1 | 2 - 3 | 504 | 112 | ||
GenBank mRNA | HY185322.1 | - | 2 - 3 | 112 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | TATA-box binding protein associated factor 4b | TAF4B |
SPGF13, TAF2C2, TAFII105 |
-33.3kb | Sense | Chr. 18 - 26226491 - 26391685 on NCBI Build GRCh38 |
Downstream | potassium channel tetramerization domain containing 1 | KCTD1 |
C18orf5, SENS |
28.9kb | Anti-sense | Chr. 18 - 26454910 - 26657401 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
