Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008264.hg.1 |
ENST00000579356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008264.hg.1 |
ENST00000579356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008264.hg.1 |
ENST00000579356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008264.hg.1 |
ENST00000581177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008264.hg.1 |
ENST00000581177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008264.hg.1 |
ENST00000581177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008264.hg.1 |
ENST00000581274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008264.hg.1 |
ENST00000581274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008264.hg.1 |
ENST00000583086.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008264.hg.1 |
ENST00000583086.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008264.hg.1 |
ENST00000583086.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008264.hg.1 |
ENST00000583086.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008264.hg.1 |
NR_110782.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008264.hg.1 |
NR_110782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156369.hg.1 |
ENST00000581274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156369.hg.1 |
ENST00000583086.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156369.hg.1 |
NR_110782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156369.hg.1 |
TC1800008264.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156369.hg.1 |
ENST00000581274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156369.hg.1 |
ENST00000583086.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156369.hg.1 |
NR_110782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008264.hg.1 |
ENST00000579356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008264.hg.1 |
ENST00000579356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008264.hg.1 |
ENST00000579356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008264.hg.1 |
ENST00000581177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008264.hg.1 |
ENST00000581177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008264.hg.1 |
ENST00000581177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008264.hg.1 |
ENST00000581274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008264.hg.1 |
ENST00000581274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008264.hg.1 |
ENST00000583086.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008264.hg.1 |
ENST00000583086.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008264.hg.1 |
ENST00000583086.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008264.hg.1 |
ENST00000583086.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008264.hg.1 |
NR_110782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008264.hg.1 |
NR_110782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156363.hg.1 |
ENST00000579356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156363.hg.1 |
ENST00000581177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156363.hg.1 |
ENST00000581274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156363.hg.1 |
NR_110782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156363.hg.1 |
TC1800008264.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156363.hg.1 |
ENST00000579356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156363.hg.1 |
ENST00000581177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156363.hg.1 |
ENST00000581274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156363.hg.1 |
NR_110782.1 |
Refseq |